Learn More
Thermo Scientific RadEye™ SPRD-GN Spectroscopic Personal Radiation Detector
Change the game in gamma and neutron radiation detection with the Thermo Scientific™ RadEye™ SPRD-GN Personal Radiation Detector
10090.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16680584
|
Thermo Scientific
4250812 |
RadEye SPRD-GN Personal Radiation Detector, includes software for configuring RadEye and Spectra download |
10090.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
Description
Get RIID capability in the palm of your hand. The Thermo Scientific™ RadEye™ SPRD-GN Personal Radiation Detector is the first pager-sized PRD that significantly exceeds the original ANSI N42-48 2008 standards for neutron alarms. With outstanding neutron performance and reliable gamma IDs, the RadEye SPRD-GN Personal Radiation Detector may be the only radiation detection tool you will need.
- Unprecedented size for gamma detection, gamma ID, and neutron detection
- Single detector enables less complex design, superior field reliability, and reduced size & weight
- Neutron alarms in <2 seconds and low false alarms exceeding ANSI specifications as well as lithium and He-3 based units
- Patent pending source-less routines for ID stabilization and outstanding neutron sensitivity
- Advanced NBR identifies artificial radiation while minimizing false alarms