Learn More
Fisherbrand™ Radiation Beta Transport Boxes
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
Brand:  Fisherbrand™ CSR-B8
Product Code. 12367127
  • 938.00 € / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Combines a shield with a convenient contained work area and storage container
  • The hinged lid creates a 15° angled shield when fully open
  • The lid can be locked shut for safety while transporting the unit and its contents
  • Side handles are fitted for easy lifting and cushioned non-slip feet prevent slipping
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes 
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
Radiation Transit Box
380 mm
75 mm
295 mm
400 mm
95 mm
315 mm
Radiation Beta Transit Box