missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rainbow Calibration Particles

Rainbow Calibration Particles are used for calibrating flow cytometers.

Brand:  Invitrogen™ A34305

Product Code. 15940443

  • 391.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The mixture of particles features:
• 8 different fluorescent intensities
• Excitation by wavelengths ranging from 365–650 nm
• Particles 3.0–3.4 μm in size

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rainbow Calibration Particle
3 to 3.4 μm
5 mL
Assorted Colors
107 particles/mL, store at 2°C to 8°C
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.