Learn More
Cytiva Rainbow™ Molecular Weight Markers
Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining. Cytiva Life Sciences™ Rainbow™ Molecular Weight Markers use bright, distinctive colors to allow easier confirmation of transfer to blotting membranes and orientation.
275.00 € - 294.00 €
Specifications
Description | Rainbow molecular weight markers enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. |
---|---|
Color | Multi |
Product Type | Protein Molecular Weight Marker/Ladder |
Form | Liquid |
Quantity | 250 μL |
Product Code | Brand | Concentration | Product Type | Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Product Type | Range | Price | Quantity & Availability | |||
10258904
|
Cytiva
RPN755E |
1 mg/mL | Protein Molecular Weight Marker/Ladder | 3500 to 38000Mr |
275.00 €
250µL |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||
10524395
|
Cytiva
RPN756E |
1.5 mg/mL | Protein Molecular Weight Marker/Ladder | 12000 to 225000Mr |
276.00 €
250µL |
Estimated Shipment: 02-10-2024
Log in to see stock. |
|||
11580684
|
Cytiva
RPN800E |
2 mg/mL | Protein Molecular Weight Marker/Ladder | 12000 to 225000Mr |
294.00 €
250µL |
In Stock
Log in to see stock. |
|||
Description
Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. The ready-to-load markers provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination.
- High intensity, prestained multicolored bands allow distinction of closely related proteins
- Full-Range Rainbow Markers (Mr 12,000 to 225,000) use ten separate proteins with six different colors
- High-Range Rainbow Markers (Mr 12,000 to 225,000) use eight separate proteins with six different colors
- Low-Range Rainbow Markers (Mr 3500 to 38,000) use seven separate proteins with five different colors
- For use on SDS-PAGE
- The bright, distinctive colors of the markers allow easier confirmation of transfer to blotting membranes and orientation.
- Markers are a mixture of individually colored proteins that are combined to produce bands of equal color intensity. Approximate protein concentration is 1 to 2 mg/mL, depending on the product
- Each 250μL pack size is sufficient for use with 50 minigels (10 × 8cm) or 25 large gels (20 × 20cm)
Specifications
Rainbow molecular weight markers enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels. | |
Protein Molecular Weight Marker/Ladder | |
250 μL | |
50 Minigels or 25 large gels |
Multi | |
Liquid | |
-15 to -30°C |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate, mesna, Glycine,N, N'-1, 2-ethanediylbis¢N-(carboxymethyl)-, sodiumsalt, hy