Learn More
Grant Instruments™ Raised Shelf for Unstirred Water Baths
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
138.00 € - 273.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11406487
|
Grant Instruments™
RS22 |
Shelf for 18 L Tanks |
202.00 €
Each |
In Stock
Log in to see stock. |
|||||
11496477
|
Grant Instruments™
RS14 |
Shelf for 12 L Tanks |
138.00 €
Each |
In Stock
Log in to see stock. |
|||||
11416487
|
Grant Instruments™
RS28 |
Shelf for 26 L Tanks |
207.00 €
Each |
In Stock
Log in to see stock. |
|||||
11426487
|
Grant Instruments™
RS38 |
Shelf for 34 L Tanks |
273.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Accommodates shallow vessels. Grant Instruments™ Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
- Choose from a variety of sizes to fit your tank.
- For use with either Stainless Steel and Plastic tank models
Specifications
Grant Heated Circulating Baths |