missing translation for 'onlineSavingsMsg'
Learn More

Raltegravir potassium salt, Thermo Scientific Chemicals

Product Code. 15741799
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871038-72-1
Molecular Formula C20H20FKN6O5
Molecular Weight (g/mol) 482.51
InChI Key IFUKBHBISRAZTF-UHFFFAOYSA-M
Synonym raltegravir potassium, raltegravir potassium salt, raltegravirpotassiumsalt, raltegravir mk-0518, potassium 4-4-fluorobenzyl carbamoyl-1-methyl-2-2-5-methyl-1,3,4-oxadiazole-2-carboxamido propan-2-yl-6-oxo-1,6-dihydropyrimidin-5-olate, isentress tn, raltegravir potassium usan:jan, pubchem22484, n-1-4-4-fluorophenyl methylcarbamoyl-5-hydroxy-1-methyl-6-oxo-pyrimidin-2-yl-1-methyl-ethyl-5-methyl-1,3,4-oxadiazole-2-carboxamide
PubChem CID 23668479
IUPAC Name potassium;4-[(4-fluorophenyl)methylcarbamoyl]-1-methyl-2-[2-[(5-methyl-1,3,4-oxadiazole-2-carbonyl)amino]propan-2-yl]-6-oxopyrimidin-5-olate
SMILES CC1=NN=C(O1)C(=O)NC(C)(C)C2=NC(=C(C(=O)N2C)[O-])C(=O)NCC3=CC=C(C=C3)F.[K+]

Specifications

Melting Point 155.0°C to 157.0°C
Quantity 250 mg
Formula Weight 482.51
Chemical Name or Material Raltegravir potassium salt
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Raltegravir potassium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Raltegravir potassium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'