missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALYL Mouse anti-Human, Unconjugated, Clone: 1F3F4, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | RALYL |
---|---|
Clone | 1F3F4 |
Concentration | 500 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880216
|
Proteintech
67534-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870216
|
Proteintech
67534-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RALYL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALYL. This antibody recognizes human, mouse, rat, pig antigen. The RALYL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
RALYL | |
500 μg/mL | |
Monoclonal | |
Mouse | |
138046 | |
Primary |
1F3F4 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG1 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto