Learn More
Thermo Scientific™ MarqMetrix Replacement Bioreactor BallProbe
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
Brand:  Thermo Scientific™ 810-02851-01
Product Code. 17749417
  • 14025.00 € / Each
Explore more special offers
Description
Description
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Bioreactor Ball Probe
MarqMetrix All-In-One Process Raman Analyzer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ MarqMetrix Replacement Bioreactor BallProbe > Replacement Part