Learn More
Thermo Scientific™ MarqMetrix Replacement Fiberhead BallProbe
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
Brand:  Thermo Scientific™ 810-02871-01
Product Code. 17769417
  • 16840.00 € / Each
Explore more special offers
Description
Description
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MarqMetrix All-In-One Process Raman Analyzer
Replacement Fiberhead BallProbe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ MarqMetrix Replacement Fiberhead BallProbe > Replacement Part