Learn More
Thermo Scientific™ MarqMetrix Replacement Process BallProbe
Replacement BallProbe for use with the MarqMetrix All-In-One process raman analyzer.
Brand:  Thermo Scientific™ 810-02841-01
Product Code. 17739417
  • 11225.00 € / Each
Explore more special offers
Description
Description
  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MarqMetrix All-In-One Process Raman Analyzer
Replacement Process BallProbe
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ MarqMetrix Replacement Process BallProbe > Replacement Part