Learn More
Thermo Scientific™ Ramsey™ Grad-Line Universal Sonic Sensor
Get dependable operation in any industrial environment with the Thermo Scientific™ Ramsey™ Grad-Line Universal Sonic Sensor.
Brand:  Thermo Scientific™ 11350
Product Code. 12141640
  • 16984.50 € / Each
Explore more special offers
Description
Description
This product has been discontinued – effective June 26, 2023. Aftermarket support including remote technical support and some field service will continue to be available as well as spare parts based on material availability.
Please check for more details on the discontinuation and contact our service team here.
At Thermo Fisher Scientific, we provide our valued customers with the products, solutions, and support that enable you to make the world healthier, cleaner, and safer.
To ensure we can continue to offer innovation, quality, value, and acceptable lead times we continuously evaluate our portfolio and take the tough decisions to discontinue products where necessary.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital
Sonic sensor
203 - 813 mm
2 Å
-20°C to +80°C (-4°F to +176°F)
0.25 turn MS, 6 pin
0.2 Å
8 - 32 in.
8 - 30 VDC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Ramsey™ Grad-Line Universal Sonic Sensor >