Learn More
OHAUS™ Ranger™ Count 4000
OHAUS™ Ranger™ Count 4000 is ideal for tough industrial counting jobs that require durability and accuracy. This scale is equipped with weighing, parts counting, check counting, and accumulation functions
Brand:  OHAUS™ 30237023
770.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15513903
  • 790.00 € / Each
Explore more special offers
Description
Description
  • Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
  • Accurate weighing and counting performance is assured with 1:1,500,000 internal counting resolution and auto-optimization software that reduces errors.
  • Easy-to-operate with SmarText™ software, a centrally located count display and dedicated Sample and Tare keys.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
210 Hours with Rechargeable Battery
Rugged Cast Aluminum Housing, Stainless Steel Platform
3 Window Backlit LCD, 3 Color Checkweighing LEDs
2.000000 g
2.000000 g
Gram, Kilogram
7.000 kg
Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately)
30.00 kg
10.000000 g
RC41M30-M
-10°C to +40°C
327 x 315 x 122 mm