Learn More
OHAUS™ Ranger™ Count 4000
OHAUS™ Ranger™ Count 4000 is ideal for tough industrial counting jobs that require durability and accuracy. This scale is equipped with weighing, parts counting, check counting, and accumulation functions
Brand: OHAUS™ 30237020
This item is not returnable.
View return policy
Description
- Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
- Accurate weighing and counting performance is assured with 1:1,500,000 internal counting resolution and auto-optimization software that reduces errors.
- Easy-to-operate with SmarText™ software, a centrally located count display and dedicated Sample and Tare keys.
Specifications
210 Hours with Rechargeable Battery | |
Rugged Cast Aluminum Housing, Stainless Steel Platform | |
3 Window Backlit LCD, 3 Color Checkweighing LEDs | |
0.200000 g | |
0.200000 g | |
Gram, Kilogram | |
7.000 kg |
Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately) | |
3.00 kg | |
1.000000 g | |
RC41M3-M | |
-10°C to +40°C | |
327 x 315 x 122 mm |