missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapamycin, 99.0%, MP Biomedicals™
Click to view available options
Quantity:
1 mg
100 μg
Specifications
Specifications
Type | Rapamycin |
Physical Form | Powder |
Assay Percent Range | 0.99 |
Formula Weight | 914.2 |
Color | White |
Quantity | 100 μg |
Chemical Name or Material | Rapamycin |
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'