missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapamycin, Fisher BioReagents
Click to view available options
Quantity:
1 mg
Packaging:
Clear Glass
Specifications
Specifications
Type | Rapamycin |
Physical Form | Powder/Solid |
CAS | 53123-88-9 |
Assay Percent Range | > 98 % |
Molecular Formula | C51H79NO13 |
Merck Index | 15, 8232 |
Packaging | Clear Glass |
InChI Key | QFJCIRLUMZQUOT-HPLJOQBZSA-N |
SMILES | CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC |
Molecular Weight (g/mol) | 914.187 |
Show More |
Safety and Handling
Product Identifier
- Rapamycin
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'