Learn More
3M™ Petrifilm™ Rapid Coliform Count Plate
Designed to provide fast, accurate results in detecting coliform contamination.
Brand:  3M™ Petrifilm™ 7100039395
Encompass
 View more versions of this product
Product Code. 17233754
  • POA / Pack of 500
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Utilizes the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of coliform.
  • Results in 4-24 hours.
  • This plate has been awarded Official Methods of Analysis (OMA) #2000.15 for Food and NF VALIDATION certificates: 3M-01/5-03/97 A for all human food products , 3M-01/5-03/97 B for All human food products and 3M-01/5-03/97 C for All human food products (except process pork products) by AFNOR Certification.
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Environmental Test
500