Learn More
3M™ Petrifilm™ Rapid E.coli / Coliform Count Plate
Povide a cost-effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples.
Brand:  3M™ Petrifilm™ 7100127433
Encompass
 View more versions of this product
Product Code. 17243784
  • 1245.21 € / Pack of 500
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • E. coli and coliform results in 18-24 hours
  • Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes.
  • Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
  • This plate has been awarded Official Methods of Analysis (OMA) #2018.13 for select food and MicroVal certificate no. 2017LR76 for a broad range of foods and select environmental surfaces by Lloyd's Register Nederland B.V. following ISO 16140-2. Food Categories tested: meat products, dairy and egg products, fish and seafood products, vegetable products, ready to eat and ready to reheat, animal feed and samples from the industrial production environment
  • Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Count Plate
Environmental Test
500