Learn More
Lonza™ Rapid Endotest - Five Research Samples
Endotoxin testing service that offers a mail-in, pre-paid services for users not requiring testing according to pharmaceutical or medical device regulations.
Brand:  Lonza™ BE95-103
Product Code. 11545789
  • 1261.82 € / Pack of 5
Explore more special offers
Description
Description
  • Endotoxin testing service for up to five research samples.
  • Samples are tested at 1:10, 1:100 and 1:1000 dilutions with a positive product control.
  • Mail-in, pre-paid service for users not requiring testing according to pharmaceutical or medical device regulations.
  • Customers can purchase Rapid Endotest packs from Lonza to submit samples to our testing service for basic determination of the endotoxin content in their sample(s).
  • Preliminary screening or inhibition/enhancement tests are not performed on samples submitted through the Pre-paid Service.
  • Unless otherwise noted, samples will be tested using the Kinetic-QCL™ LAL Assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not according to pharmaceutical or medical device regulations.
Mail-in, pre-paid testing service.
Endotoxin Testing Service
Endotoxin testing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lonza™ Rapid Endotest - Five Research Samples > Mail-in, pre-paid testing service.