missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rapid Equilibrium Dialysis (RED) Inserts and Plates

Equilibrium dialysis is a method used in plasma protein binding (PPB) studies to determine the amount of free, active drugs present in plasma. The Rapid Equilibrium Dialysis (RED) Device is widely accepted as the gold standard device for measuring interactions between drugs and PPB.

Brand:  Thermo Scientific™ 90087

 View more versions of this product

Product Code. 11776456

  • 233.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store at room temperature
Cellulose Membrane, Dialysis
10 Inserts
Competition RED Device Base Plate
Binding Equilibrium, Protein
12 kDa
Equilibrium Dialysis
Pierce
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI4442493 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number90087
Lot NumberYI4040517 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number90087
Lot NumberYF3965446 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog Number90087
Lot NumberYD3842981 Certificate TypeCertificate of Analysis Date08/04/2023 Catalog Number90087
Lot NumberVL3159661 Certificate TypeCertificate of Analysis Date16/01/2021 Catalog Number90087
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Rapid Equilibrium Dialysis (RED) Inserts and Plates > Pierce Competition RED Device Inserts, 12K MWCO; 10 Inserts

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Patent pending.