missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit
Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit is two-component, high-precision, detergent-compatible assay optimized to measure (at 480 nm) total protein concentration compared to protein standard curve of known concentrations.
48.28 € - 261.00 €
Specifications
Assay Range | 20 to 2000 μg/mL |
---|---|
Content And Storage | Store in refrigerator (2–8°C). |
Assay | Rapid Gold BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Alert:
Store at 4°C
Description
- A fast, two-component, high-precision, detergent-compatible assay optimized to determine total protein concentration
- Can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications
- Assay maintains the key characteristics of the traditional BCA assay but allows a fast time and room temperature incubation equal to dye-binding methods
- Provides comparable accuracy but with a 5 min., room temperature incubation and measured at 480 nm
- This improvement eliminates the need to wait or expose the samples to elevated temperatures for a fast time to results
- Exhibits similar protein-to-protein variation as traditional BCA Protein Assay and less protein-to-protein variation than dye-binding protein assay methods (Bradford)
- Unaffected by typical concentrations of most ionic and non-ionic detergents
- Linear working range for BSA of 20 to 2000 μg/mL
- Store in refrigerator at 2°C to 8°C
Specifications
20 to 2000 μg/mL | |
Rapid Gold BCA Assay | |
Pierce™ | |
Not Target-Specific | |
Spectrophotometer, Microplate Reader |
Store in refrigerator (2–8°C). | |
Colorimetric | |
Protein Quantitation Assay | |
Solution-based Detection, Absorbance | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title