Learn More
Thermo Scientific™ RapID™ NF PLUS System
Identify over 70 medically important, oxidase-positive, gram-negative bacilli, including Vibrio spp. in just four hours using this rapid identifcation system.
Brand: Thermo Scientific™ R8311005
Description
Identify over 70 medically important, oxidase-positive, gram-negative bacilli, including Vibrio spp. in just four hours using the Thermo Scientific™ RapID™ NF PLUS System, an identification system based on enzyme technology.
The RapID NF PLUS System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important glucose-nonfermenting, Gram-negative bacteria and other select glucose-fermenting, Gram-negative bacteria not belonging to the family Enterobacteriaceae, which have been isolated from human clincal specimens.- One-step inoculation—decreased prep time for increased productivity
- Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
- Four-hour incubation*—quicker time-to-result for a faster response
- Common procedure—lessened materials usage saves time and money
- Visible color reactions—reduced subjectivity decreases repeat testing
- Better reporting—more coverage, advanced updates
A clear plastic tray contains reagent impregnated wells, allowing simultaneuos inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Innoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.
After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a databse or through the use of a computer-generated Code Compedium (ERIC).
Requires:
RapID Spot Indole, (R8309002); RapID Nitrate A (R8309003); RapID Inoculation Fluid, (R8325102)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 8°C | |
Yes | |
1 Box |
RapID NF PLUS System | |
Gram-negative bacilli (Oxidase-positive) | |
20 Panels/Kit |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6202861 | Certificate of Analysis | 28/02/2025 | R8311005 |
6198998 | Certificate of Analysis | 06/01/2025 | R8311005 |
6154294 | Certificate of Analysis | 25/11/2024 | R8311005 |
6139396 | Certificate of Analysis | 04/11/2024 | R8311005 |
6176470 | Certificate of Analysis | 19/10/2024 | R8311005 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.