missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ QC Set

Obtain faster recovery, enhanced viability, and direct-streak-to-plate capabilities with QC organisms manufactured using an unmatched proprietary gel preservation process.

615.00 € - 1360.00 €

Specifications

Certifications/Compliance ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
Content And Storage 2°C to 8°C
Format Culti-Loops
Target Organism Class Organism Sets
Test Type Qualitative QC
View More Specs

Products 7
Product Code Brand Description Product Type Price Quantity & Availability  
Product Code Brand Description Product Type Price Quantity & Availability  
12995973
View Documents
Thermo Scientific™
R4653050
RapID™ ANA II QC Set/3 Bacteroides uniformis, Clostridium sordellii, Parabacteroides distasonis
1040.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12905983
View Documents
Thermo Scientific™
R4653051
RapID™ NH QC Set/4 Haemophilus influenzae Type a, Aggregatibacter aphrophilus, Moraxella catarrhalis, Oligella urethralis
1270.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12975973
View Documents
Thermo Scientific™
R4653047
RapID™ STAPH PLUS QC Set/4 Enterobacter aerogenes, Oligella ureolytica, Staphylococcus haemolyticus, Staphylococcus saprophyticus
755.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12925983
View Documents
Thermo Scientific™
R4653053
RapID™ SS/u QC Set/5 Enterococcus faecalis, Escherichia coli, Klebsiella pneumoniae subsp. pneumoniae, Proteus mirabilis, Serratia marcescens
980.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12978244
View Documents
Thermo Scientific™
R4653054
RapID™ NF PLUS QC Set/4 Acinetobacter baumannii, Aeromonas hydrophila, Elizabethkingia meningoseptica, Oligella ureolytica
615.00 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
12955983
View Documents
Thermo Scientific™
R4653060
RapID™ YEAST PLUS QC Set/5 Candida albicans, Candida glabrata, Candida kefyr, Cryptococcus laurentii, Yarrowia lipolytica
1360.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12945983
View Documents
Thermo Scientific™
R4653056
RapID™ ONE QC Set/4 Enterobacter aerogenes, Escherichia coli, Proteus vulgaris, Pseudomonas aeruginosa
805.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ensure high-performance products, reproducible clinical test results and consistent microbiological evaluation of pharmaceuticals and food with Thermo Scientific™ Culti-Loops™ quality control organisms. Thermo Scientific™ RapID™ QC Sets include a ready-to-use set of QC organisms in Culti-Loops format which are ready-to-use, disposable bacteriological loops containing stabilized, viable microorganisms in a unique gel matrix.

Trust Thermo Scientific quality control organisms for quality results you can depend on, backed by over 30 years of expertise in the manufacturing and packaging of controlled microorganisms.

Convenience
  • Decrease time and unnecessary costs of laboratory resources with a direct-streak inoculating loop that requires no rehydration.
Performance
  • Obtain enhanced viability and recovery with proprietary gel-preserved organisms.


Value
  • Consolidate ordering time and shipping costs by turning to one supplier for high-quality culture media and quality control organisms.
Confidence
  • Reach complete traceability with ATCC™ License Derivative strains.


Efficiency
  • Achieve accurate and convenient documentation with peel-off, transferable labels.


Safety
  • Reduce the risk of contamination and infection with zero organism handling.




  • More than 600 quality control organisms, including convenient sets for commonly used identification systems
  • ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
  • ATCC™ Licensed Derivative Program member


Not all products are available for sale in all territories. Please inquire.


Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.


*The ATCC Licensed Derivative™ Emblem, the ATCC Licensed Derivative™ word mark, and the ATCC catalog marks are trademarks of ATCC™. Thermo Fisher Scientific is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications

Specifications

ISO certified, cGMP (current Good Manufacturing Practices) compliant, FDA registered
Culti-Loops
Qualitative QC
2°C to 8°C
Organism Sets
Videos
SDS
Documents

Documents

Product Certifications