Learn More
3M™ Petrifilm™ Rapid Yeast and Mold Count Plate
Simplify and accelerate detection. Featuring a new indicator technology that makes colonies easier to interpret, these completely sample-ready plates return results in just 48 hours.
Brand:  3M™ Petrifilm™ 7100040141
Encompass
Product Code. 17213764
Description
Description
  • Utilizes the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of yeast and mold.
  • Results in 48 hours.
  • Proven as reliable as DRBC and DG-18 spread plate methods, as well as acidified PDA and YGC agar pour plate methods.
  • Can be used with both low and high water activity foods.
  • Eliminates the need for the labor intensive, multiple plate spread plate technique.
  • Versatility for air, swab or surface contact environmental sampling.
Specifications
Specifications
Environmental Test
500