Learn More
Labconco™ RapidVap Vacuum Dry Evaporation System
The RapidVap Vacuum Evaporation Systems are ideal for preparation of samples in a variety of applications including drug discovery, agrichemistry, mycology testing and environmental analysis.
Brand:  Labconco™ 7900001
Product Code. 11761549
  • 16205.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The RapidVap Vacuum Evaporation Systems are ideal for preparation of samples in a variety of applications including drug discovery, agrichemistry, mycology testing and environmental analysis. The systems use vacuum along with heat and vortex motion to help speed evaporation and provide an alternative to traditional centrifugal evaporation methods. These systems may be used with a broad range of aggressive chemicals. Heat is supplied by a dry block heating system that, unlike water baths, adds no potential source of contamination and requires no maintenance.
Specifications
Specifications
0 to 300 V
PTFE-coated Aluminum
1000-watt dry block heating system
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products