missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Itgam, Rat, Cy5, Clone: M1/70, Abnova™
Rat monoclonal antibody raised against Itgam.
Brand: Abnova MAB9866.100ug
This item is not returnable.
View return policy
Specifications
Itgam | |
Monoclonal | |
Cyanine5 | |
Flow Cytometry (≤ 0.3 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Itgam | |
Itgam | |
Native Itgam from mouse splenic T cells and splenocytes. | |
RUO | |
16409 | |
Store at 4°C. Do not freeze. | |
IgG2b κ |
Flow Cytometry | |
M1/70 | |
Rat monoclonal antibody raised against Itgam. | |
In PBS (0.09% sodium azide) | |
CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction