Learn More
Invitrogen™ Rat anti-Mouse IgG (H+L) Secondary Antibody, HRP
Rat Monoclonal Secondary Antibody
Brand: Invitrogen™ 046020
433.35 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
This monoclonal Rat anti-Mouse IgG cocktail reacts with all subclasses of mouse IgG and exhibits minimal reactivity to mouse IgA, IgM, and light chains. Cross-reactivity has been observed with swine and human IgG.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Mouse IgG (H+L) | |
Monoclonal | |
1 mg/mL | |
PBS with 4mg/mL BSA, 40% glycerol and 0.19% Kathon™ CG; pH 7.4 | |
Gamma Immunoglobin. | |
1 mL | |
Secondary | |
4°C | |
Liquid |
Western Blot, Immunocytochemistry, ELISA | |
LO-MG-7, LO-MG-1-2, and LO-MG3-7 | |
HRP | |
Rat | |
purified | |
RUO | |
Mouse | |
Secondary Antibody | |
IgG1 κ |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.