missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Anti-Mouse IgG1, AKP, Clone: X56, BD
Brand: BD Biosciences 557272
This item is not returnable.
View return policy
Description
The X56 antibody reacts specifically with mouse IgG1 of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: X56
Isotype: IgG1 κ
Species Reactivity: Mouse
ELISA
Specifications
IgG1 | |
X56 | |
Affinity Purified | |
RUO | |
Secondary |
Monoclonal | |
Alkaline Phosphatase (AP) | |
1 mL | |
Immunology | |
Mouse |