Learn More
R&D Systems™ Rat C-Reactive Protein/CRP DuoSet ELISA
Rat C-Reactive Protein/CRP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DY1744
Product Code. 30021605
  • 947.00 € / 1 set
Explore more special offers
Description
Description
The Rat C-Reactive Protein/CRP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rat C-Reactive Protein/CRP.
Specifications
Specifications
Biotin
ELISA
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
156.00 to 10000 pg/mL
25419