Learn More
Invitrogen™ Rat LMAN1 (aa 159-173) Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP277
Product Code. 11544900
  • 163.77 € / 35µg
Explore more special offers
Description
Description
Rat ERGIC-53 (residues 159-173) Synthetic Peptide, PEP-277, from Invitrogen.
PEP-277 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 159-173 of rat ERGIC-53. The sequence of this peptide is (amino to carboxy terminus): F (159)-D-S-F-D-N-D-G-K-K-N-N-P-A-I (173).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, ERGIC-53, catalog # PA1-074. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
116666
purified
Rat LMAN1 (aa 159-173)
-20°C. Avoid freeze-thaw cycles.
Lyophilized
Control
Yes
Peptide
RUO - research use only
Rat
Q62902
0.35 mg/mL
Synthetic Peptide
peptide with no preservative
Lman1
No preservative
35 μg
F(159) - D - S - F - D - N - D - G - K - K - N - N - P - A - I(173)