Learn More
Novus Biologicals™ Rat Multi-tissue Tissue MicroArray (Normal)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78074
Product Code. 18632841
  • 385.00 € / Pack of 5
Explore more special offers
Specifications
5 Slides
Rat
Please bake slides at 60C for 30 minutes before use.