missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598346
This item is not returnable.
View return policy
Description
RB38 is a protein coding gene and its exact function is unknown.
Specifications
RB38 | |
Polyclonal | |
Unconjugated | |
RB38 | |
Rabbit | |
Protein G | |
RUO | |
5728724 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2.96 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
RB38 | |
Recombinant Chlamydomonas reinhardtii 38 kDa RNA-binding protein (1-382AA) | |
100 μL | |
Primary | |
Algae | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RB38 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction