missing translation for 'onlineSavingsMsg'
Learn More

RBS™ Alkaline Cleaning Agent

Product Code. 12863512
Click to view available options
Product Line:
RBS 25
RBS 35
RBS 50
Unit Size:
20 L
30kg
Each
This item is not returnable. View return policy

Product Code. 12863512

Brand: RBS RBS50DRUM

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for Soaking and Cleaning Surfaces and Radioactive Decontamination

  • RB525 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking
  • RB535 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking and permits lengthy cleaning without calcium precipitation
  • RB550 is non-foaming, with bacterial action with a pH of 10.6 when using 10g per L of water, suitable for soaking and automatic washing machines
  • All three are suitable for ultrasonic cleaners
  • RBSVIRO is a neutral foaming, surfacing, non-toxic, non-irritant and non-corrosive agent with a pH of 7, suitable for soaking
Recommended for use in research, pathology, chemistry, biology, food and drink, pharmaceutical, electronics, optical and glass industry laboratories.

Specifications

Product Type Alkaline Cleaning Agent
Product Line RBS 50
Appearance Colorless
For Use With (Application) Suitable for Soaking and Automatic Washing Machines
Quantity 30 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.