Learn More
RBS™Neutral Detergent
RBS, for manual cleaning, soaking and ultrasonic cleaning of sensitive glassware, metals and plastics
Brand: RBS RBST230
This item is not returnable.
View return policy
Description
RBS, for manual cleaning, soaking and ultrasonic cleaning of sensitive glassware, metals and plastics Medline ScientificSpecifications
Neutral Detergent | |
5 L |
Product Content Correction
Product Title
RBS™Neutral Detergent >
Spot an opportunity for improvement?Share a Content Correction