Learn More
Hirschmann™ rea-stabil™ Liquid Bottle
Helps avoid algae, bacteria and mould formation in the laboratory. Hirschmann Instruments™ rea-stabil™ Liquid Bottle is for use with hot water baths, thermostats and similar areas of use.
Brand: Hirschmann™ 9780100
This item is not returnable.
View return policy
Description
- Bactericide and fungicide
- Non-toxic
- Very economical - 0.1% working concentration
Specifications
Stabilization Concentrate | |
Liquid | |
0.1% | |
Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- rea-stabil
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.