Learn More
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators
Use these gas regulators to deliver the right amount of gas for sample evaporation using the Thermo Scientific™ ReactiVap™ Evaporator.
Brand:  Thermo Scientific™ 60181-625
345.37 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15224613
  • 347.10 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Deliver the right amount of gas to your Thermo Scientific™ ReactiVap™ Evaporator using these gas regulators. The Reacti-Vap Evaporator delivers pressurized gas to easily and efficiently evaporate samples for gas chromatography (GC) analysis. Choose between gas regulators with a 1/8 inch or a 1/4 inch tube fitting.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas Regulator, 1/8in. Tube Fitting
Reacti-Vap Evaporators
Gas Regulator (1/8 in. Tube Fitting)
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.