Learn More
Thermo Scientific™ Reacti-Vap™ Evaporator PTFE-Coated Needles
Use these needles to deliver gas and evaporate samples on the Thermo Scientific™ Reacti-Vap™ Evaporator.
Brand:  Thermo Scientific™ TS-18784
 View more versions of this product
Product Code. 11507490
  • 217.08 € / Pack of 9
Explore more special offers
Description
Description
Use Thermo Scientific™ Reacti-Vap™ PTFE-Coated Needles to deliver gas and evaporate samples on Reacti-Vap Evaporators. These blunt-ended, 19-gauge, stainless steel needles reduce cross-contamination and corrosion when evaporating solvents that contain strong acids. The needles were specifically designed for Reacti-Vap Evaporators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE-coated
19 G
Syringe Needle
Reacti-Vap Evaporators
9 Pack
4 in.
102 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.