missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Reaction Buffer for phi29 DNA Polymerase (10X)
Thermo Scientific 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.
Brand: Thermo Scientific™ B62
This item is not returnable.
View return policy
Description
Thermo Scientific 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.
Specifications
Reaction Buffer | |
5 x 1 mL | |
• 10X Reaction buffer for Phi29 DNA Polymerase (5 x 1.0 mL) |
|
10X |
Isothermal Amplification | |
Traditional Cloning | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
User Guide: 10X Reaction Buffer for phi29 Polymerase
Version: A.0
File size: 75.3 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Reaction Buffer for phi29 DNA Polymerase (10X) > 5 x 1 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction