missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Reactive Blue 4, dye content min. 35%

45.83 € - 121.61 €

Chemical Identifiers

CAS 13324-20-4
Molecular Formula C23H14Cl2N6O8S2
Molecular Weight (g/mol) 637.42
MDL Number MFCD00001218
InChI Key RTLULCVBFCRQKI-UHFFFAOYSA-N
Synonym C.I. 61205; Procion™ Blue MX-R
PubChem CID 25863
IUPAC Name 1-amino-4-[3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid
SMILES NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459723
View Documents
Thermo Scientific Chemicals
A12911.14
25 g
45.83 €
25g
In Stock
to see stock.
Add to basket
 
11469723
View Documents
Thermo Scientific Chemicals
A12911.22
100 g
121.61 €
100g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13324-20-4
637.42
RTLULCVBFCRQKI-UHFFFAOYSA-N
25863
NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
C23H14Cl2N6O8S2
MFCD00001218
C.I. 61205; Procion™ Blue MX-R
1-amino-4-[3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid

Specifications

13324-20-4
MFCD00001218
RTLULCVBFCRQKI-UHFFFAOYSA-N
1-amino-4-[3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid
25863
Odorless
C23H14Cl2N6O8S2
C.I. 61205; Procion™ Blue MX-R
NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
637.42
637.44
Reactive Blue 4, dye content ≈40%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H335

missing translation for 'einecsNumber' : 236-363-9

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.