Learn More
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
Brand:  Cytiva 25-6603-96
 View more versions of this product
Product Code. 12997160
  • 1682.00 € / Each
Explore more special offers
Description
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Genomic DNA Amplification
96
> 10 kb
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
1 x 2.6mL 2X Denaturation Buffer, 3 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit 96 Reactions
4 h