missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Reagent F-Actin Phalloidin Conjugates

Perfect for fixed cells and tissue, ReadyProbes Reagent F-actin phalloidin conjugates offer clear visualization of actin filaments, making it perfect for cell biology, neuroscience, cancer research, and more.

Brand:  Invitrogen™ R37110

198.66 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 14879680

  • 301.00 € / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 2 x 2.5 mL dropper bottles
This item is not returnable. View return policy

Description

Description

ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is a room temperature-stable solution of Alexa Fluor 488 Phalloidin (Cat. No. A12379) packaged in a dropper bottle. ActinRed 555 ReadyProbes Reagent is a comparable F-actin stain conjugated to the red-orange fluorescent dye tetramethylrhodamine (TRITC). ActinRed 555 reagent is a room temperature-stable solution of Rhodamine Phalloidin (Cat. No. R415) packaged in a dropper bottle.

These ready-to-use solutions combines superior brightness with exceptional F-actin staining. Phalloidin is a bi-cyclic peptide, commonly used in imaging applications to selectively label F-actin. Fluorescently labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals and low non-specific binding.

Features

  • High-affinity staining of F-actin with superior specificity compared to antibody methods
  • ActinGreen 488 Excitation/Emission: (495/518 nm), detected through standard GFP and FITC filter sets
  • ActinRed 555 Excitation/Emission: (540/565 nm), detected through standard RFP and TRITC filters sets
  • Use counterstain (i.e., DAPI or NucBlue ReadyProbes reagent) as desired

ReadyProbes format advantages

  • Ready-to-use formulation—no need to dilute, weigh, or pipette
  • Just use two drops per mL of medium to stain, incubate 30 minutes, wash, then image
  • Stable at room temperature—keep handy on the benchtop

Applications

  • For fixed and permeabilized cells or tissue
  • Fluorescence microscopy (IF/IHC/ICC)
  • High content screening, high content imaging
  • Flow cytometry

Suggestions for use

  • Both ActinRed 555 and ActinGreen 488 reagents may be added directly to fixed cells in full medium or buffer solutions
  • In most cases 2 drops/mL and an incubation time of 15–30 minutes is sufficient for bright actin staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
495/518
Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Fluorescence
Visible
ActinGreen™, ReadyProbes™
Fluorescent Dye
Cytoskeleton

2 x 2.5 mL dropper bottles

Store at ≤ 25°C.

Alexa Fluor™ 488
1 kit
ActinGreen™ 488 ReadyProbes™ Reagent (AlexaFluor™ 488 phalloidin)
Liquid
Fluorescence Intensity
Phalloidin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.