missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Reagent F-Actin Phalloidin Conjugates

Perfect for fixed cells and tissue, ReadyProbes Reagent F-actin phalloidin conjugates offer clear visualization of actin filaments, making it perfect for cell biology, neuroscience, cancer research, and more.

Brand:  Invitrogen™ R37112

 View more versions of this product

Product Code. 15119325

  • 310.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: 2 x 2.5 mL dropper bottles
This item is not returnable. View return policy

Description

Description

ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is a room temperature-stable solution of Alexa Fluor 488 Phalloidin (Cat. No. A12379) packaged in a dropper bottle. ActinRed 555 ReadyProbes Reagent is a comparable F-actin stain conjugated to the red-orange fluorescent dye tetramethylrhodamine (TRITC). ActinRed 555 reagent is a room temperature-stable solution of Rhodamine Phalloidin (Cat. No. R415) packaged in a dropper bottle.

These ready-to-use solutions combines superior brightness with exceptional F-actin staining. Phalloidin is a bi-cyclic peptide, commonly used in imaging applications to selectively label F-actin. Fluorescently labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals and low non-specific binding.

Features

  • High-affinity staining of F-actin with superior specificity compared to antibody methods
  • ActinGreen 488 Excitation/Emission: (495/518 nm), detected through standard GFP and FITC filter sets
  • ActinRed 555 Excitation/Emission: (540/565 nm), detected through standard RFP and TRITC filters sets
  • Use counterstain (i.e., DAPI or NucBlue ReadyProbes reagent) as desired

ReadyProbes format advantages

  • Ready-to-use formulation—no need to dilute, weigh, or pipette
  • Just use two drops per mL of medium to stain, incubate 30 minutes, wash, then image
  • Stable at room temperature—keep handy on the benchtop

Applications

  • For fixed and permeabilized cells or tissue
  • Fluorescence microscopy (IF/IHC/ICC)
  • High content screening, high content imaging
  • Flow cytometry

Suggestions for use

  • Both ActinRed 555 and ActinGreen 488 reagents may be added directly to fixed cells in full medium or buffer solutions
  • In most cases 2 drops/mL and an incubation time of 15–30 minutes is sufficient for bright actin staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Red-Orange
540/565
Confocal Microscope, EVOS™, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Fluorescence
Visible
ActinRed™, ReadyProbes™
Fluorescent Dye
Cytoskeleton

2 x 2.5 mL dropper bottles

Store at ≤ 25°C.

TRITC (Tetramethylrhodamine Isothiocyanate)
2 dropper bottle(s) kit
ActinRed™ 555 ReadyProbes™ Reagent (Rhodamine phalloidin)
Liquid
Fluorescence Intensity
Phalloidin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.