missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagent Alcohol, CHROMASOLV™, for HPLC, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Specifications
Specifications
Absorbance | max. 0.35 at 220 nm, max. 0.65 at 210 nm, max. 1.00 at 205 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 255 nm |
CAS | 64-17-5, 67-56-1, 67-63-0 |
CAS Max % | 90.0000%, 5.0000%, 5.0000% |
Density | 0.7847 g/cm3 |
Boiling Point | 78.32°C |
Quantity | 1 L |
Flash Point | 15°C |
Packaging | Glass Bottle |
Molecular Formula | C2H6O |
MDL Number | MFCD00003568 |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'