missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Reagent Bottle, Wide Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck Product Code.: 11746958
Add to basket

DWK Life Sciences Reagent Bottle, Wide Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck

Product Code. 11746958 Shop All DWK Life Sciences Products
1000 mL
Click to view available options
Capacity (Metric):
50 mL
100 mL
500 mL
1000 mL
This item is not returnable. View return policy

Product Code. 11746958

Brand: DWK Life Sciences 231855401

Add to basket
Add to basket
This item is not returnable. View return policy

Classic wide-neck, glass-stoppered reagent soda lime glass bottles.

A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!

  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All glass components
  • Tight closure due to standard ground joint
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules
Includes With standard ground glass flat-headed stopper
Material Soda Lime Glass
Quantity 10/Pk.
Autoclavable Not Autoclavable
Diameter (Metric) 107 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Reagent Bottle, Wide Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.