Learn More
BD Biosciences Reagent Rack - PE
Pre-cooled reagent rack keeps your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability.
Brand: BD Biosciences 657178
This item is not returnable.
View return policy
Description
Pre-cooled BD™ Reagent Racks keep your antibodies cooled for 75 minutes, preserving their quality and protein-dye stability. The BD Reagent Racks are stackable and available in different colors to help you keep your vials organized on the bench as well as in the fridge. Each rack is filled with cooling material, and can accommodate up to 25 BD™ Amber vials and 20 narrower BD Pharmingen™ type vials. Size 180 mm x 54,5 mm - Weight: 950 gr.Specifications
Filled with cooling material. | |
Yes | |
950 G |
BD™ Amber Vials (5) and Narrower BD Pharmingen™ Type Vials (20) | |
180 mm x 54.5 mm |