missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Reagent Reservoir, Non-sterile PVC
BIOPHARMAGREENER_CHOICE

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures

68.30 € - 76.35 €

Specifications

Certifications/Compliance Dnase, Rnase & Pyrogen Free
Coating None
Color Transparent
Description Bulk wrap
Material PVC
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16384862
BIOPHARMAGREENER_CHOICE
View Documents
Heathrow Scientific™
120690
50 mL
68.30 €
Pack of 100
Estimated Shipment: 06-03-2025
to see stock.
Add to basket
 
16354862
BIOPHARMAGREENER_CHOICE
View Documents
Heathrow Scientific™
120650
100 mL
76.35 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
  • Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Quality certificate available on request
  • Disposable
  • Design Patent Pending
Specifications

Specifications

Dnase, Rnase & Pyrogen Free
Transparent
PVC
Reagent Reservoir
Reservoir
40°C Operating
100
Not Autoclavable
Single-use
None
Bulk wrap
Non-sterile
Reagent
104°F Operating
600
100
1
Conical
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.