missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene



Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures
40.65 € - 237.00 €
Specifications
Certifications/Compliance | Dnase, Rnase & Pyrogen Free |
---|---|
Coating | None |
Sterility | Sterile |
Type | Reagent Reservoir |
For Use With (Application) | Reagent |
Product Code | Brand | Capacity (Metric) | Packaging | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Packaging | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
16334852
![]() ![]() |
Heathrow Scientific™
120637 |
5 mL | 1/bag, 50 bags | 2.8 x 0.8 x 0.7 in |
40.65 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||
16354852
![]() ![]() |
Heathrow Scientific™
120639 |
10 mL | 1/bag, 100 bags | 2.8 x 0.8 x 0.7 in |
82.50 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
16364852
![]() ![]() |
Heathrow Scientific™
120640 |
25 mL | 5/bag, 40 bags | 4 x 1 x 0.8 in |
237.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
16374852
![]() ![]() |
Heathrow Scientific™
120641 |
25 mL | 1/bag, 50 bags | 4 x 1 x 0.8 in |
131.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
16314862
![]() ![]() |
Heathrow Scientific™
120646 |
50 mL | 5/bag, 20 bags | 4 x 1.1 x 1.2 in |
122.57 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
16394852
![]() ![]() |
Heathrow Scientific™
120643 |
50 mL | 5/bag, 40 bags | 4 x 1.1 x 1.2 in |
184.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
16317701
![]() ![]() |
Heathrow Scientific™
120644 |
50 mL | 1/bag, 100 bags | 4 x 1.1 x 1.2 in |
118.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
16324862
![]() ![]() |
Heathrow Scientific™
120647 |
100 mL | 5/bag, 40 bags | 4.3 x 1.9 x 1.3 in |
196.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
16334862
![]() |
Heathrow Scientific™
120648 |
100 mL | 1/bag, 100 bags | 4.3 x 1.9 x 1.3 in |
179.00 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
Description
- Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
- Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Quality certificate available on request
- Disposable
- Design Patent Pending
Specifications
Dnase, Rnase & Pyrogen Free | |
Sterile | |
Reagent | |
104°F Operating | |
Not Autoclavable | |
Single-use |
None | |
Reagent Reservoir | |
Reservoir | |
40°C Operating | |
1 | |
Conical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene