Learn More
Thermo Scientific™ Reagent Reservoirs

Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
Brand: Thermo Scientific™ 95128095
160.00 EUR valid until 2025-05-31
Use promo code "25076" to get your promotional price.
Description
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Features:
- Trough within a trough maximizes the amount of liquid accessible to pipette tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
- 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent

Specifications
White | |
Polystyrene | |
Reagent Reservoir with Divider | |
Single-use |
25 mL | |
Non-sterile | |
10 Bags of 10 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2422212 | Certificate of Compliance | 06/11/2024 | 95128095 |
2335612 | Certificate of Compliance | 17/10/2024 | 95128095 |
2414312 | Certificate of Compliance | 17/10/2024 | 95128095 |
2327912 | Certificate of Compliance | 06/02/2024 | 95128095 |
2315112 | Certificate of Compliance | 04/09/2023 | 95128095 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.