Learn More
Heidolph™ Reax 2
Compact shaker for small volumes.
Brand:  Heidolph™ 541-21001-06
Product Code. 17343020
  • 1305.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Reax 2 with the optional adapter for test tubes is designed for simultaneous mixing of up to 20 vessels with diameters of 10 to 18 mm. This corresponds to the size of classic test tubes or 15 mL Falcon tubes. Alternatively, you can insert your complete rack into the universal adapter (width: 230 mm, height min./max.: 45 mm/160 mm) of the overhead shaker.
  • Equipped with a solid metal frame, its design stands for extreme load capacity and reliability. Guaranteed safety during unattended continuous operation.
Specifications
Specifications
230 V
UK Plug