missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Reax 2 with Universal Adapter
Compact shaker with adapter for test tubes.
Brand: Heidolph™ 541-21009-03
1591.25 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 6.76000kg
Description
- Quickly clamps in 2 vessels of any size and accepts load capacities up to 1 kg
- Speed continuously adjustable from 20 to 100 rpm
- Universal adaptor allows for the use of various vessels of 50 to 160 mm height
- Includes adapter for the use of 20 test tubes at a time
Specifications
230 V | |
UK Plug |