Learn More
Heidolph™ Reax Control with Small Pick-Up Attachment
Reax Control is a precise model with speed scaling.
Brand:  Heidolph™ 541-11000-04
Product Code. 17303020
  • 503.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All the outstanding features of the Reax Top can be found in this model: a rotational speed of up to 2500 rpm and a 5 mm shaking orbit
  • The electronic speed control and the scaling of the speed display allow precise settings
  • Applications that require a determined speed are thus shaken more gently
  • Shear force-sensitive organisms remain particularly protected by the constant speed in the low range
Specifications
Specifications
230 V
UK Plug